Learn More
MSC Insize Steel Rule, Stainless Steel, 150 mm, 6 in, ±0.18 mm Accuracy
Catalog No. 5000536247
Encompass
This rigid steel rule from Insize is crafted from stainless steel, offering durability and a non-glare surface for easy reading. It provides a measuring range of 150 mm (6 in) with an accuracy of ±0.18 mm, suitable for precise measurements in various technical and scientific applications.
  • Made of stainless steel for durability
  • Rigid construction ensures straight and accurate measurements
  • Non-glare surface for clear and easy reading
  • Features graduation in 0.5 mm, 1 mm, 1/64 in, 1/32 in, and 1/16 in
  • Metric graduation displayed on the front side
  • Inch graduation displayed on the back side
Catalog No. 50-005-36247 Supplier MSC Supplier No. 15522337
This rigid steel rule from Insize is crafted from stainless steel, offering durability and a non-glare surface for easy reading. It provides a measuring range of 150 mm (6 in) with an accuracy of ±0.18 mm, suitable for precise measurements in various technical and scientific applications.
  • Made of stainless steel for durability
  • Rigid construction ensures straight and accurate measurements
  • Non-glare surface for clear and easy reading
  • Features graduation in 0.5 mm, 1 mm, 1/64 in, 1/32 in, and 1/16 in
  • Metric graduation displayed on the front side
  • Inch graduation displayed on the back side