Learn More
MSC Insize USA LLC Digital Protractor, 0-360° Range, 0.05° Resolution, 10 in Blade
Catalog No. 5000529088
Encompass
The Insize USA LLC Digital Protractor is an electronic tool designed for precise angle measurements. It offers a full 0-360° measuring range with high resolution and accuracy, making it suitable for a variety of industrial and laboratory applications.
  • On/off, hold, zero, and ABS buttons for absolute and incremental measurement
  • Locking screws allow fixation at 4 positions for specific measuring ranges, or removal for a full 360° range
  • Comes with a protective case
  • Features automatic power off to conserve battery life
  • Includes a level vial for accurate positioning
  • Powered by a CR2032 battery
Catalog No. 50-005-29088 Supplier MSC Supplier No. 19068816
The Insize USA LLC Digital Protractor is an electronic tool designed for precise angle measurements. It offers a full 0-360° measuring range with high resolution and accuracy, making it suitable for a variety of industrial and laboratory applications.
  • On/off, hold, zero, and ABS buttons for absolute and incremental measurement
  • Locking screws allow fixation at 4 positions for specific measuring ranges, or removal for a full 360° range
  • Comes with a protective case
  • Features automatic power off to conserve battery life
  • Includes a level vial for accurate positioning
  • Powered by a CR2032 battery