Learn More
MSC Insize USA LLC Electronic Caliper, 0-200 mm, 0.01 mm, Left Hand
Catalog No. 5000532327
Encompass
This electronic caliper is designed for precise measurements in various applications. Made of stainless steel, it offers a measuring range of 0-200 mm (0-8 in) with a resolution of 0.01 mm (0.0005 in). This tool features data output capabilities and is specifically designed for left-hand users.
  • Designed for left-hand users.
  • Automatic power off; power turns on by moving the digital unit.
  • Measures inside diameter, step, outside diameter, and depth.
  • Knife-edge jaw type.
  • Thumb wheel jaw adjustment.
  • Powered by one CR2032 lithium battery.
  • Includes manufacturer inspection certificate and hard plastic case.
  • Complies with DIN862 standards.
  • Data output is compatible with SPC cable or wireless transmitter.
Catalog No. 50-005-32327 Supplier MSC Supplier No. 18458281
This electronic caliper is designed for precise measurements in various applications. Made of stainless steel, it offers a measuring range of 0-200 mm (0-8 in) with a resolution of 0.01 mm (0.0005 in). This tool features data output capabilities and is specifically designed for left-hand users.
  • Designed for left-hand users.
  • Automatic power off; power turns on by moving the digital unit.
  • Measures inside diameter, step, outside diameter, and depth.
  • Knife-edge jaw type.
  • Thumb wheel jaw adjustment.
  • Powered by one CR2032 lithium battery.
  • Includes manufacturer inspection certificate and hard plastic case.
  • Complies with DIN862 standards.
  • Data output is compatible with SPC cable or wireless transmitter.