Learn More
MSC Insize USA LLC Electronic Caliper, 0-300 mm, 0.01 mm Resolution, Data Output, Stainless Steel
Catalog No. 5000527116
Encompass
This stainless steel electronic caliper is designed for versatile inside, outside, step, and depth measurements. It provides a 0-300 mm (0-12 in) range with ±0.01 mm (±0.0005 in) resolution and ±0.03 mm (±0.0015 in) accuracy. Features data output for SPC/Wireless Transmitter, CR2032 battery power, and a NIST-traceable inspection certificate. It's designed for left-hand users and comes with a hard plastic case.
  • Left-hand user design.
  • Dual resolution: .0005"/0.01mm.
  • 0-12 inches (0-300mm) measuring range.
  • On/off, zero, inch/mm conversion buttons.
  • Automatic power off; power on by moving digital unit.
  • Durable stainless steel.
  • Data output capable.
  • Includes NIST-traceable inspection certificate.
Catalog No. 50-005-27116 Supplier MSC Supplier No. 18459255
This stainless steel electronic caliper is designed for versatile inside, outside, step, and depth measurements. It provides a 0-300 mm (0-12 in) range with ±0.01 mm (±0.0005 in) resolution and ±0.03 mm (±0.0015 in) accuracy. Features data output for SPC/Wireless Transmitter, CR2032 battery power, and a NIST-traceable inspection certificate. It's designed for left-hand users and comes with a hard plastic case.
  • Left-hand user design.
  • Dual resolution: .0005"/0.01mm.
  • 0-12 inches (0-300mm) measuring range.
  • On/off, zero, inch/mm conversion buttons.
  • Automatic power off; power on by moving digital unit.
  • Durable stainless steel.
  • Data output capable.
  • Includes NIST-traceable inspection certificate.