Learn More
MSC Insize USA LLC Electronic Caliper, 150 mm, 0.01 mm Resolution, Plastic
Catalog No. 5000536342
Encompass
This electronic caliper offers a maximum measurement range of 150 mm or 6 inches with a resolution of ±0.01 mm. Constructed from hard plastic, both the caliper body and jaws are non-magnetic and non-electrically conductive, which helps prevent scratching of workpieces. It is suitable for Inside Diameter, Step, Outside Diameter, and Depth measurements.
  • Maximum measurement: 150 mm (6 in)
  • Resolution: ±0.01 mm (±0.0005 in)
  • Accuracy: ±0.10 mm (±0.0040 in)
  • Plastic construction for caliper body and jaws
  • Inside/outside jaw style with knife-edge type
  • Functions for inside diameter, step, outside diameter, and depth measurements
  • Powered by one CR2032 lithium battery
  • Features on/off, zero, and inch/mm buttons
  • Automatic power-off function
  • Includes inside diameter jaws and a depth measurement rod
  • Comes with a hard plastic case
Catalog No. 50-005-36342 Supplier MSC Supplier No. 15630379
This electronic caliper offers a maximum measurement range of 150 mm or 6 inches with a resolution of ±0.01 mm. Constructed from hard plastic, both the caliper body and jaws are non-magnetic and non-electrically conductive, which helps prevent scratching of workpieces. It is suitable for Inside Diameter, Step, Outside Diameter, and Depth measurements.
  • Maximum measurement: 150 mm (6 in)
  • Resolution: ±0.01 mm (±0.0005 in)
  • Accuracy: ±0.10 mm (±0.0040 in)
  • Plastic construction for caliper body and jaws
  • Inside/outside jaw style with knife-edge type
  • Functions for inside diameter, step, outside diameter, and depth measurements
  • Powered by one CR2032 lithium battery
  • Features on/off, zero, and inch/mm buttons
  • Automatic power-off function
  • Includes inside diameter jaws and a depth measurement rod
  • Comes with a hard plastic case