Learn More
MSC Insize USA LLC Electronic Caliper, 150 mm Range, 0.01 mm Resolution, Stainless Steel
Catalog No. 5000513623
Encompass
This electronic caliper is designed for precise measurements in professional and industrial settings. It offers a broad measurement range and high resolution, making it suitable for demanding applications, particularly for measuring grooves inside bores. Constructed from durable stainless steel, it ensures reliability and longevity.
  • Measures up to 150 mm (6 in)
  • Features 0.01 mm (0.0005 in) resolution
  • Provides data output capabilities for easy data transfer
  • Made from stainless steel for enhanced durability
  • Specifically designed for groove measurements inside bores
  • Includes a hard plastic case for protection
  • Equipped with automatic power off to conserve battery
  • Offset jaw for specialized measurements
Catalog No. 50-005-13623 Supplier MSC Supplier No. 19108315
This electronic caliper is designed for precise measurements in professional and industrial settings. It offers a broad measurement range and high resolution, making it suitable for demanding applications, particularly for measuring grooves inside bores. Constructed from durable stainless steel, it ensures reliability and longevity.
  • Measures up to 150 mm (6 in)
  • Features 0.01 mm (0.0005 in) resolution
  • Provides data output capabilities for easy data transfer
  • Made from stainless steel for enhanced durability
  • Specifically designed for groove measurements inside bores
  • Includes a hard plastic case for protection
  • Equipped with automatic power off to conserve battery
  • Offset jaw for specialized measurements