Learn More
MSC Insize USA LLC Electronic Caliper, 300 mm, 0.01 mm Resolution, Stainless Steel
Catalog No. 5000513348
Encompass
This electronic caliper is designed for precise measurements, offering a maximum measurement of 300 mm with a 0.01 mm resolution. Constructed from durable stainless steel, it is suitable for various applications including inside diameter, outside diameter, depth, and step measurements, and features data output capabilities.
  • Data output available
  • Constructed from stainless steel
  • Includes inspection certificate traceable to NIST
  • Features on/off, zero, and inch/mm conversion buttons
  • Automatic power off with activation by movement
  • Comes with a depth measurement rod
  • Supplied with manufacturer inspection certificate
  • Packaged in a hard plastic case
  • Performs depth, inside diameter, outside diameter, and step measurements
  • Powered by a CR2032 lithium battery
  • Conforms to DIN862 standards
Catalog No. 50-005-13348 Supplier MSC Supplier No. 28730059
This electronic caliper is designed for precise measurements, offering a maximum measurement of 300 mm with a 0.01 mm resolution. Constructed from durable stainless steel, it is suitable for various applications including inside diameter, outside diameter, depth, and step measurements, and features data output capabilities.
  • Data output available
  • Constructed from stainless steel
  • Includes inspection certificate traceable to NIST
  • Features on/off, zero, and inch/mm conversion buttons
  • Automatic power off with activation by movement
  • Comes with a depth measurement rod
  • Supplied with manufacturer inspection certificate
  • Packaged in a hard plastic case
  • Performs depth, inside diameter, outside diameter, and step measurements
  • Powered by a CR2032 lithium battery
  • Conforms to DIN862 standards