Learn More
MSC Insize USA LLC Electronic Depth Gage, 0-6 in/0-150 mm, 0.0005 in/0.01 mm Resolution
Catalog No. 5000536344
Encompass
This electronic depth gage is designed to provide quality and innovative measurements. It is intended to meet or exceed all international standards. Key features include automatic power off and the ability to power on by moving the digital unit, enhancing user convenience. It also comes with a hard plastic case for protection.
  • On/Off, Zero, Inch/mm buttons
  • Automatic power off
  • Digital unit activates power when moved
  • Includes a hard plastic case
  • Optional data output cable available
  • Optional extension base available
Catalog No. 50-005-36344 Supplier MSC Supplier No. 14967988
This electronic depth gage is designed to provide quality and innovative measurements. It is intended to meet or exceed all international standards. Key features include automatic power off and the ability to power on by moving the digital unit, enhancing user convenience. It also comes with a hard plastic case for protection.
  • On/Off, Zero, Inch/mm buttons
  • Automatic power off
  • Digital unit activates power when moved
  • Includes a hard plastic case
  • Optional data output cable available
  • Optional extension base available