Learn More
MSC Insize USA LLC Mechanical Outside Micrometer, 3 in Range, 0.0001 in Graduation, Solid Carbide Measuring Faces
Catalog No. 5000513524
Encompass
This mechanical outside micrometer offers precise measurements with a 3 inch range and 0.0001 inch graduation. It features a ratchet stop thimble and a rotating spindle, ensuring reliable and consistent results. The micrometer is calibrated and equipped with solid carbide measuring faces, and it comes with a manufacturer inspection certificate traceable to NIST, along with a protective hard plastic case.
  • Maximum measurement of 3.0000 inch
  • Graduation of 0.000100 inch
  • Ratchet stop thimble for consistent pressure
  • Calibrated for accuracy
  • Rotating spindle for precise adjustments
  • Solid carbide measuring faces for durability
  • Supplied with manufacturer inspection certificate traceable to NIST
  • Includes a protective case
  • Accuracy of 0.00020 inch (for 2-3 inch range)
Catalog No. 50-005-13524 Supplier MSC Supplier No. 18111245
This mechanical outside micrometer offers precise measurements with a 3 inch range and 0.0001 inch graduation. It features a ratchet stop thimble and a rotating spindle, ensuring reliable and consistent results. The micrometer is calibrated and equipped with solid carbide measuring faces, and it comes with a manufacturer inspection certificate traceable to NIST, along with a protective hard plastic case.
  • Maximum measurement of 3.0000 inch
  • Graduation of 0.000100 inch
  • Ratchet stop thimble for consistent pressure
  • Calibrated for accuracy
  • Rotating spindle for precise adjustments
  • Solid carbide measuring faces for durability
  • Supplied with manufacturer inspection certificate traceable to NIST
  • Includes a protective case
  • Accuracy of 0.00020 inch (for 2-3 inch range)