Learn More
MSC Box Partners Cool Barrier Insulated Box Liner, 14x10x10 in., 25/Pack
Catalog No. 501919835
Encompass
These leak resistant liners are designed to create insulated shipping containers. They are one-piece gusseted, set up quickly, and are made with FDA compliant materials suitable for incidental food contact. Use with cold packs to maintain product quality, taste, texture, and color during transit.
  • Leak resistant
  • Create insulated shipping containers
  • One-piece gusseted design
  • Quick and easy setup
  • Made with FDA compliant materials for incidental food contact
  • Ensures product quality, taste, texture, and color when used with cold packs
Catalog No. 50-191-9835 Supplier MSC Supplier No. 39594544
These leak resistant liners are designed to create insulated shipping containers. They are one-piece gusseted, set up quickly, and are made with FDA compliant materials suitable for incidental food contact. Use with cold packs to maintain product quality, taste, texture, and color during transit.
  • Leak resistant
  • Create insulated shipping containers
  • One-piece gusseted design
  • Quick and easy setup
  • Made with FDA compliant materials for incidental food contact
  • Ensures product quality, taste, texture, and color when used with cold packs