Learn More
MSC Value Collection Box Liner, 24x18x18in, Silver, 1 ea
Catalog No. 501919836
Encompass
This box liner is designed to create an insulated shipping container for various products. It features a leak-resistant design and is suitable for maintaining product quality when used with cold packs.
  • One-piece gusseted liner design allows for quick and easy setup
  • FDA approved for incidental contact with food
  • Can be used with cold packs to maintain product quality, taste, texture, and color
  • Creates an insulated shipping container
Catalog No. 50-191-9836 Supplier MSC Supplier No. 39594551
This box liner is designed to create an insulated shipping container for various products. It features a leak-resistant design and is suitable for maintaining product quality when used with cold packs.
  • One-piece gusseted liner design allows for quick and easy setup
  • FDA approved for incidental contact with food
  • Can be used with cold packs to maintain product quality, taste, texture, and color
  • Creates an insulated shipping container