Learn More
MSC Partners Brand Insulated Box Liners, 12x12x12 in, Silver, 25/Pk.
Catalog No. 501917761
Encompass
Insulated box liners are designed to maintain temperature for shipping temperature-sensitive items, providing protection against external temperature fluctuations. These liners are suitable for various packaging needs.
  • Constructed with aluminum foil exterior and 3/16 in bubble lining for effective insulation
  • Measures 12 in long, 12 in wide, and 12 in high
  • Features a silver color
  • Supplied in a package quantity of 25
Catalog No. 50-191-7761 Supplier MSC Supplier No. 39573407
Insulated box liners are designed to maintain temperature for shipping temperature-sensitive items, providing protection against external temperature fluctuations. These liners are suitable for various packaging needs.
  • Constructed with aluminum foil exterior and 3/16 in bubble lining for effective insulation
  • Measures 12 in long, 12 in wide, and 12 in high
  • Features a silver color
  • Supplied in a package quantity of 25