Learn More
MSC Polar Tech Industries Insulated Container, 12x12x11-1/2 in, 1 ea
Catalog No. 501920725
Encompass
This insulated shipping container is designed for the safe transport of temperature-sensitive biological and chemical materials. It includes a bio foam container and a cardboard shipping carton, providing thermal protection for 1-5 days.
  • Made from bio foam, cardboard, and styrofoam
  • White color for easy identification
  • Suitable for perishable shipping applications
  • Includes both foam container and shipping carton
  • Features a 2 in wall thickness for insulation
  • Inside dimensions are 12 x 12 x 11-1/2 in
Catalog No. 50-192-0725 Supplier MSC Supplier No. 39559281
This insulated shipping container is designed for the safe transport of temperature-sensitive biological and chemical materials. It includes a bio foam container and a cardboard shipping carton, providing thermal protection for 1-5 days.
  • Made from bio foam, cardboard, and styrofoam
  • White color for easy identification
  • Suitable for perishable shipping applications
  • Includes both foam container and shipping carton
  • Features a 2 in wall thickness for insulation
  • Inside dimensions are 12 x 12 x 11-1/2 in