Learn More
Aviva Systems Biology Corp INSM1 Antibody - middle region (ARP31990_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31990_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against INSM1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human INSM1. Alternate names: IA1, IA-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52849