Learn More
Aviva Systems Biology Corp INSM1 Antibody - C-terminal region (ARP32667_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32667_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INSM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human INSM1. Alternate names: IA1, IA-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-54698