Learn More
Aviva Systems Biology Corp INSM2 Antibody - middle region (ARP30029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INSM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human INSM2. Alternate names: IA-6, mlt1, IA6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54104