Learn More
BioVendor, LLC Sircol; INSOLUBLE Collagen Assay, Bovine Collagen (Large Economy Kit Size: 400 assays), Multispecies colorimetric assay, Biocolor Item # S6000
SDP
Catalog No. 501725965
Encompass
Sircol; INSOLUBLE Collagen Assay, Bovine Collagen (Large Economy Kit Size: 400 assays). The Sircol; INSOLUBLE Collagen Assay is a dye-binding method for analysis of insoluble collagen fibers. The assay be used to assess the rate of production of newly laid down collagen fibers during periods of rapid growth, development, tissue repair, remodeling and wound healing. Sources of material includes tissues, bone and calcified tissue. It replaces the conventional hydroxyproline analysis. No more working with concentrated acids or alkalis! The Sircol Insoluble Assay uses mild acid and temperature treatment for 2 to 3 hrs. The solubility conversion to denatured collagen and subsequent measurement can be completed within one working day. The ratio of soluble and insoluble collagens can be measured using the same Sircol Dye Reagent. Multispecies colorimetric assay. Detection Limit: 10.0 g. Assay-time: 4 hours. Biocolor Item # S6000
Catalog No. 50-172-5965 Supplier BioVendor, LLC Supplier No. S6000
Sircol; INSOLUBLE Collagen Assay, Bovine Collagen (Large Economy Kit Size: 400 assays). The Sircol; INSOLUBLE Collagen Assay is a dye-binding method for analysis of insoluble collagen fibers. The assay be used to assess the rate of production of newly laid down collagen fibers during periods of rapid growth, development, tissue repair, remodeling and wound healing. Sources of material includes tissues, bone and calcified tissue. It replaces the conventional hydroxyproline analysis. No more working with concentrated acids or alkalis! The Sircol Insoluble Assay uses mild acid and temperature treatment for 2 to 3 hrs. The solubility conversion to denatured collagen and subsequent measurement can be completed within one working day. The ratio of soluble and insoluble collagens can be measured using the same Sircol Dye Reagent. Multispecies colorimetric assay. Detection Limit: 10.0 g. Assay-time: 4 hours. Biocolor Item # S6000