Learn More
Advanced Biomatrix Bovine Collagen, Type I, Lyophilized Fibrous Powder, 1 gram
Catalog No. 501960282
Encompass_Preferred
Type I bovine collagen, lyophilized fibrous powder, Catalog Number 5162, is extracted from bovine flexor tendon with the raw material sourced from closed/controlled herds of animals. Because the collagen is extracted from tendon, the material is more naturally crosslinked. The collagen is polymeric insoluble collagen. This product is supplied as a lyophilized fibrous powder in a 1 gram package size. Bioburden and endotoxin levels are tested; this product is not considered sterile. This collagen product is naturally cross-linked yielding a robust material for applications which require structure and strength. This product can be readily prepared into such forms as tissue scaffolds, foams, sponges, suspensions, coatings, putties, films and sheets but does not form hydrogels.
Catalog No. 50-196-0282 Supplier Advanced Biomatrix Supplier No. 51621GM
Type I bovine collagen, lyophilized fibrous powder, Catalog Number 5162, is extracted from bovine flexor tendon with the raw material sourced from closed/controlled herds of animals. Because the collagen is extracted from tendon, the material is more naturally crosslinked. The collagen is polymeric insoluble collagen. This product is supplied as a lyophilized fibrous powder in a 1 gram package size. Bioburden and endotoxin levels are tested; this product is not considered sterile. This collagen product is naturally cross-linked yielding a robust material for applications which require structure and strength. This product can be readily prepared into such forms as tissue scaffolds, foams, sponges, suspensions, coatings, putties, films and sheets but does not form hydrogels.