Learn More
Sigma Aldrich Fine Chemicals Biosciences InSolution Cytochalasin D
Catalog No. 502899447
Encompass
A cell-permeable fungal toxin that is approximately 10 times more potent than cytochalasin B (Cat. No. 250233) in inhibiting actin filament function. Does not inhibit monosaccharide transport across cell membranes. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments at 10 181M. Inactivates low conductance K channels. Also modulates CD4 cross-linking in T lymphocytes and increases intracellular Ca2 levels. Cytochalasin D also exhibits antibiotic and anti-tumor properties.
Catalog No. 50-289-9447 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5047760001
A cell-permeable fungal toxin that is approximately 10 times more potent than cytochalasin B (Cat. No. 250233) in inhibiting actin filament function. Does not inhibit monosaccharide transport across cell membranes. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments at 10 181M. Inactivates low conductance K channels. Also modulates CD4 cross-linking in T lymphocytes and increases intracellular Ca2 levels. Cytochalasin D also exhibits antibiotic and anti-tumor properties.