Learn More
UNITED SCOPE LLC DBA AMSCOPE AmScope SM-1B Series Inspection and Dissecting Binocular Zoom Stereo Microscope 7X-45X Magnification with 64 LED Compact Ring Light and 5MP USB 3.0 C-mount Camera on Compact Pillar Stand
SDP
Catalog No. NC3769411
Encompass_Preferred
7X-45X zoom magnification using high-quality Greenough stereoscopic objective lenses.Continuous zoom magnification. Super widefield optics offer crystal clear images with a large field-of-view. Sturdy pillar stand with a small footprint. Monitor and capture photos and videos with the included 5MP USB 3.0 camera. Professional microscopy software for Windows, Mac, and Linux provide tools for image processing, measuring, and more.The 64-LED ring-light produces an encompassing field of bright light with reduced shadows for use with any application.
Catalog No. NC3769411 Supplier UNITED SCOPE LLC DBA AMSCOPE Supplier No. SM1BS64S5M3
7X-45X zoom magnification using high-quality Greenough stereoscopic objective lenses.Continuous zoom magnification. Super widefield optics offer crystal clear images with a large field-of-view. Sturdy pillar stand with a small footprint. Monitor and capture photos and videos with the included 5MP USB 3.0 camera. Professional microscopy software for Windows, Mac, and Linux provide tools for image processing, measuring, and more.The 64-LED ring-light produces an encompassing field of bright light with reduced shadows for use with any application.