Learn More
Fritsch Milling And Sizing Inc Inspection Certificate 3.1B En 10204 Analysette 3 Pro
SDP
Catalog No. 501534721
Encompass
Calibration certificate 3.1 EN 10204 for FRITSCH Sieve Shakers ANALYSETTE 3 PRO
The inspection certificate 3.1 EN 10204 enables the use of the Sieve Shaker analysette 3 PRO as calibration and validation inspection instrument.
Catalog No. 50-153-4721 Supplier Fritsch Milling And Sizing Inc Supplier No. 96.0010.00
Calibration certificate 3.1 EN 10204 for FRITSCH Sieve Shakers ANALYSETTE 3 PRO
The inspection certificate 3.1 EN 10204 enables the use of the Sieve Shaker analysette 3 PRO as calibration and validation inspection instrument.