Learn More
Micronova™ Inspection Mirror with Plastic Bottom Plate
SDP
Catalog No. 19116696 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Material:
Stainless Steel, Polypropylene
Catalog No. Material
19-116-696 Stainless Steel, Polypropylene
1 options
Catalog No. 19-116-696 Supplier Micronova™ Supplier No. QDIST66P
Add to Cart
Add to Cart
An electropolished stainless steel mirror with a swivel joint and handle connection for inspecting hard to reach areas.
  • A highly polished stainless steel mirror for inspecting hard to reach areas
  • Features a swivel connection allowing it to be attached to fixed and extension handles
  • This allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned
  • The optional flashlight attachment is ideal for use in tanks and other low light applications
  • QD versions connect with SSU series stainless steel handles
  • Certifications
    Certificate of conformance
    Specifications
    Product Type Snap-on Inspection Mirror
    Sterility Non-sterile
    Autoclavable Autoclavable
    Color-coded No
    Material Stainless Steel, Polypropylene