Learn More
Grainger Inspection Mirror Round SS 10 to 37
Catalog No. 502766233
Encompass_Preferred
The Ullman Inspection Mirror is a 2-3/8 inch diameter mirror with a telescoping handle that extends from 17 to 29 inches. The mirror has a 2x magnification for close inspection work. The handle is chrome-plated steel and has a cushion grip. The mirror tilts and swivels for optimal viewing angles. This high quality inspection mirror is ideal for automotive, industrial, and home use.
Catalog No. 50-276-6233 Supplier Grainger Supplier No. 16W188
The Ullman Inspection Mirror is a 2-3/8 inch diameter mirror with a telescoping handle that extends from 17 to 29 inches. The mirror has a 2x magnification for close inspection work. The handle is chrome-plated steel and has a cushion grip. The mirror tilts and swivels for optimal viewing angles. This high quality inspection mirror is ideal for automotive, industrial, and home use.