Learn More
Grainger InspectionMirror Rnd SS 11 1/8 -30 1/4
Catalog No. 502766382
Encompass_Preferred
The Ullman 3-3/8" Inspection Mirror is a chrome-plated steel mirror with a 3-3/8" diameter. It has a telescoping handle that extends from 7" to 15" for inspecting hard-to-reach places. The mirror provides a clear, distortion-free reflection. It can be used for auto repair, plumbing, electrical work, HVAC, and machinery inspection. This durable inspection mirror is ideal for mechanics, maintenance workers, and do-it-yourselfers.
Catalog No. 50-276-6382 Supplier Grainger Supplier No. 16W189
The Ullman 3-3/8" Inspection Mirror is a chrome-plated steel mirror with a 3-3/8" diameter. It has a telescoping handle that extends from 7" to 15" for inspecting hard-to-reach places. The mirror provides a clear, distortion-free reflection. It can be used for auto repair, plumbing, electrical work, HVAC, and machinery inspection. This durable inspection mirror is ideal for mechanics, maintenance workers, and do-it-yourselfers.