Learn More
Com-Pac International™ Inspex™ Self-Standing Specimen Bag
Catalog No. 22043307
Change view
Click to view available options
Dimensions (L x W):
6 x 9in.
6 x 12in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-043-307 6 x 12in.
22-043-306 6 x 9in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-043-307 Supplier Com-Pac International™ Supplier No. 9722P
Add to Cart
Add to Cart
This item is not returnable. View return policy
Clear, leak-proof, resealable. The Inspex™ Self Standing Specimen Bags stand up all by themselves.
  • Leak-tight double zipper and side seals
  • Super clear, high barrier film contains odors and resists punctures
  • Bottom gusset enables bag to remain standing while being filled and specimen is inserted or retrieved
  • Bottom gusset allows bags to be stored upright
  • Rear document pouch
Specifications
Autoclavable Not Autoclavable
Closure Type Zipper
Color Clear
No. per Case 1000 (4 packs)
No. per Pack 250
Dimensions (L x W) 6 x 12in.
Length (English) 6 in.