Learn More
Insphero Inc InSphero™ Incubox - powered by InSphero
SDP
Catalog No. NC2216387
Encompass
The Incubox™ is a specially engineered container which creates a high-humidity microclimate inside conventional cell culture incubators and protects your sensitive plates from large humidity drops and fluctuations – the major cause for evaporation effects.1. Maintain a stable medium volume and accurate compound concentration due to minimal evaporation over time2. Reduce well-to-well variation by minimizing evaporation-induced reagent gradients across the plate3. Use all wells of your plate by eliminating edge effects in your culture plate4. Save on plasticware by using up to 38% more wells of your plate1 Incubox™, water bath, venting holes for continuous gas exchange; built of transparent, inert and temperature stable material, non-sterile.For further information, please visit www.insphero.com
This item is not returnable. View return policy
Catalog No. NC2216387 Supplier Insphero Inc Supplier No. CSAH11
The Incubox™ is a specially engineered container which creates a high-humidity microclimate inside conventional cell culture incubators and protects your sensitive plates from large humidity drops and fluctuations – the major cause for evaporation effects.1. Maintain a stable medium volume and accurate compound concentration due to minimal evaporation over time2. Reduce well-to-well variation by minimizing evaporation-induced reagent gradients across the plate3. Use all wells of your plate by eliminating edge effects in your culture plate4. Save on plasticware by using up to 38% more wells of your plate1 Incubox™, water bath, venting holes for continuous gas exchange; built of transparent, inert and temperature stable material, non-sterile.For further information, please visit www.insphero.com