Learn More
Enzo Life Sciences Insulin receptor (1142-1153) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP3140500
Encompass_Preferred
This sequence, TRDIYETDYYRK, is from the insulin receptor β-subunit cytoplasmic domain, inclusive of regulatory autophosphorylation sites Tyr1146, Tyr1150 and Tyr1151. It has been used as an insulin receptor tyrosine kinase substrate and the phosphorylated peptide as a protein tyrosine phosphatase substrate. Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1341