Learn More
Carnabio Usa Inc INSR/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 081425
Encompass
INSR, Catalytic domain, Wild type, Amino Acid 1005-1310, NP_000199.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0467