Learn More
Sino Biological Recombinant Human IKK beta Protein (GST Tag), Active
Catalog No. 502574667
Encompass_Preferred
Recombinant human IKKβ (1-662) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4667 Supplier Sino Biological Supplier No. I0310BG50
Recombinant human IKKβ (1-662) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.