Learn More
Enzo Life Sciences Insulin receptor (pTyr1158/1162/1163) and IGF-1 Receptor (pTyr1131/1135/1136) polyclonal antibody (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLSA3920020
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of human insulin receptor phosphorylated at Tyr1158, Tyr1162 and Tyr1163. Species reactivity: Human, Mouse. Specificity: Recognizes insulin receptor/IGF-1R phosphorylated at Tyr1158/1162/1163. Applications: WB. Formulation: Liquid. In PBS, pH 7.4, containing BSA and sodium azide.
Catalog No. 50-201-1809