Learn More
Aviva Systems Biology Corp INSR Antibody - middle region (ARP41374_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41374_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INSR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human INSR. Alternate names: CD220, HHF5ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57826