Learn More
Aviva Systems Biology Corp Insr Antibody - middle region (ARP41373_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41373_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Insr. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-284-930