Learn More
Enzo Life Sciences Insulin receptor beta subunit monoclonal antibody (CT-3) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA4320100
Encompass_Preferred
Clone: CT3. Isotype: IgG1. Immunogen: C-terminal 100 amino acids of human insulin receptor β subunit. Species reactivity: Human, Mouse, Rat, Monkey. Specificity: Recognizes the β subunit of the insulin receptor. Applications: IHC, WB. Formulation: Liquid. In 10 mM PBS, pH 7.4, containing 0.2% BSA, and 0.09% sodium azide.
Catalog No. 50-201-1823