Learn More
Enzo Life Sciences Insulin receptor beta monoclonal antibody (C18C4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905683100
Encompass_Preferred
Clone: C18C4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant cytoplasmic domain of human Insulin Receptor β. UniProt ID: P06213. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Hamster, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-200-6991