Learn More
Enzo Life Sciences [pTyr1322]Insulin receptor monoclonal antibody (21G12) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905646100
Encompass_Preferred
Clone: 21G12. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of insulin receptor phosphorylated at Tyr1322. UniProt ID: P06213. Species reactivity: Human, Mouse, Dog. Applications: ELISA, WB. Recommended Dilutions: ELISA (0.1µg/ml) Western Blot (0.5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-6981