Learn More
Trajan Scientific L-MARK Brand Insert With bottom Spring 250 l, 6x29, Case of 1000
Catalog No. 501251121
Encompass_Preferred
L-MARK Brand, Insert, With bottom Spring, 250 uL, Clear, 6x29mm, Case of 1000, For use With Large Opening 2mL Vials. Made in the USA.
Catalog No. 50-125-1121 Supplier Trajan Scientific Supplier No. LPP.2005010
L-MARK Brand, Insert, With bottom Spring, 250 uL, Clear, 6x29mm, Case of 1000, For use With Large Opening 2mL Vials. Made in the USA.