Learn More
World Precision Instrument Sterilization Instrument Cassettes with Spring Holding Lids-WP-4244
SDP
Catalog No. 502090193
Encompass
  • Manufactured with stainless steel 304
  • Electropolished
  • Smooth rounded corners
This item is not returnable. View return policy
Catalog No. 50-209-0193 Supplier World Precision Instrument Supplier No. WP4244
  • Manufactured with stainless steel 304
  • Electropolished
  • Smooth rounded corners