Learn More
New England Biolabs, Inc. Instant Sticky-end Ligase Master Mix – 50 reactions
SDP
Catalog No. NC0412453
Encompass
Instant Sticky-end Ligase Master Mix is a ready-to-use 2X solution of T4 DNA ligase and a proprietary ligation enhancer in an optimized reaction buffer. Specifically formulated to rapidly ligate cohesive-end (2-4 bp) substrates and improve transformation, the mix simplifies reaction set-up and provides an optimized ratio of enzyme and buffer components. No thawing of the master mix is required as it maintains a liquid state during storage at -20C and no incubation time is necessary to achieve ligation efficiencies sufficient for successful sub-cloning of sticky-end substrates. Just add the master mix to DNA with compatible ends, mix and transform; thereby reducing the valuable time needed for routine ligations. Ligations for subcloning can be carried out in small volumes with low concentrations, allowing users to conserve precious DNA samples, and be used directly to transform many strains of chemically competent E. coli without dilution.
Catalog No. NC0412453 Supplier New England Biolabs, Inc. Supplier No. M0370S
Instant Sticky-end Ligase Master Mix is a ready-to-use 2X solution of T4 DNA ligase and a proprietary ligation enhancer in an optimized reaction buffer. Specifically formulated to rapidly ligate cohesive-end (2-4 bp) substrates and improve transformation, the mix simplifies reaction set-up and provides an optimized ratio of enzyme and buffer components. No thawing of the master mix is required as it maintains a liquid state during storage at -20C and no incubation time is necessary to achieve ligation efficiencies sufficient for successful sub-cloning of sticky-end substrates. Just add the master mix to DNA with compatible ends, mix and transform; thereby reducing the valuable time needed for routine ligations. Ligations for subcloning can be carried out in small volumes with low concentrations, allowing users to conserve precious DNA samples, and be used directly to transform many strains of chemically competent E. coli without dilution.