Learn More
Roboz Surgical INST TRAY 12.25X7.75X2.25IN
SDP
Catalog No. NC9564143
Encompass
Solid Instrument Tray; 12 X 7.75 X 2.25 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles A
Catalog No. NC9564143 Supplier Roboz Surgical Supplier No. RT1402S
Solid Instrument Tray; 12 X 7.75 X 2.25 in. These Trays Are Electro-Polished To Provide A Tough, Corrosion-Resistant Surface And A Bright, Clean Look.Constructed Of Heavy-Duty, 304 18-8 Stainless Steel, Pmp Trays Are Available In Many Different Styles A