Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Essential Lab Insta-Chek pH Test Strip Test Range 0 to 14

Catalog No. 13641145 Shop All Micro Essential Lab Products
Change view
Click to view available options
pH Range:
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
13-641-145 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-641-145 Supplier Micro Essential Lab Supplier No. 80014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conveniently packed in flip-top vials, these multipad pH strips provide excellent color separation over the entire pH scale with clear multicolor matches at every interval.

Conveniently packed in flip-top vials, these premium quality multipad pH strips provide excellent color separation over the entire pH scale.

  • Clear, bright multicolor matches at every whole interval from pH 0 to 14
  • Color chart has pH matches at 0, 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, and 14
  • Case of 6 vials. (Each vial contains 100 strips)

Specifications

pH Range 0 to 14
Quantity Case of 6 vials. (Each vial contains 100 strips.)
Range Wide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.