Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insta-Chek™ Wide-Range Paper Dispenser
SCI_ED

Catalog No. S60149A Shop All Micro Essential Lab Products
Change view
Click to view available options
Type:
Paper Dispenser
pH Test Strip Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S60149A pH Test Strip Refill
S60149 Paper Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S60149A Supplier Micro Essential Lab Supplier No. JR113/EMD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Single color match for each pH value.

  • 14 distinct colors match the 14 pH values for accurate readings
  • Includes color chart
  • Dispenser contains 50 ft. × 0.5 in.W roll (15.2m × 1.27cm)

Specifications

pH Range 0 to 13
For Use With (Application) For accurate reading
Includes Color chart
Quantity Each
Type pH Test Strip Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.