Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Insta-Test 6-Way Drinking Water Test Strip
SDPSCI_ED

Catalog No. S43421 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
25 Test Strips/Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S43421 25 Test Strips/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43421 Supplier LaMotte™ Supplier No. 2933G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With a single dip, the strip tests chlorine, pH, hardness, nitrate and nitrite in less than 45 seconds.

Range (ppm):

  • Chlorine (Free and Total): 0 to 10
  • pH: 4 to 10
  • Hardness: 0 to 400
  • Nitrate: 0 to 50
  • Nitrite: 0 to 10

Sensitivity (ppm):

  • Chlorine (Free and Total): 0,0.5, 1, 3, 5, 10
  • pH: 4, 5, 6, 7, 8, 9, 10
  • Hardness: 0, 50, 100, 200, 400
  • Nitrate: 0, 5, 10, 25, 50 (NO3-N)
  • Nitrite: 0, 0.5, 1.9, 5.0, 10.0 (NO2-N)

Specifications

Color Indicator Waterproof color chart on the label
Format Packaged in pop-top vial with integrated desiccant liner
For Use With (Application) Municipal and well water testing
Quantity 25 Test Strips/Bottle
Test Time <45 sec.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.