Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Insta-Test™ Peracetic Acid Single Factor Test Strip
SDPSCI_ED

Catalog No. S43598 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
50 Strips/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S43598 50 Strips/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43598 Supplier LaMotte™ Supplier No. 3000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A convenient way to quickly test water for peracetic acid.

  • Range: 0, 10, 20, 50, 85, 160ppm
  • 50 Tests

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Peracetic acid
Quantity 50 Strips/Vial
Range 0-10-20-40-60-85-160 ppm
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Type Single Factor Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.