Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
SDPSCI_ED

Catalog No. S72315 Shop All LaMotte Company Products
Change view
Click to view available options
Parameter Specifications:
Chlorine - free, high range
Chlorine - free, low range
Chlorine - total, low range
Chlorine dioxide
Copper
Peracetic Acid
Sodium Chloride
Quantity:
10 Papers/Blister Card, 10 Cards/Pk.
25 Strips/Vial
50 Strips/Vial
Range:
0 to 10 ppm
0 to 10 ppm (0, 0.25, 0.5, 1, 3, 10)
0 to 50 ppm
0 to 500 ppm
0-0.3-0.6-1-3 ppm
0-0.5-1-3-5-10 ppm
0-50-100-250-500-800 ppm
1500 to 5000 ppm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Parameter Specifications Quantity Range
S72315 Chlorine - free, high range 50 Strips/Vial 0-50-100-250-500-800 ppm
S72408 Chlorine dioxide 50 Strips/Vial 0 to 10 ppm
S72409 Chlorine dioxide 50 Strips/Vial 0 to 500 ppm
S72410 Peracetic Acid 50 Strips/Vial 0 to 50 ppm
S72328 Chlorine - free, low range 25 Strips/Vial 0-0.5-1-3-5-10 ppm
S72330 Copper 25 Strips/Vial 0-0.3-0.6-1-3 ppm
S72331 Sodium Chloride 10 Papers/Blister Card, 10 Cards/Pk. 1500 to 5000 ppm
S72329 Chlorine - total, low range 25 Strips/Vial 0 to 10 ppm (0, 0.25, 0.5, 1, 3, 10)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. S72315 Supplier LaMotte™ Supplier No. 3031
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For water quality testing.

  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time
  • Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †

Specifications

For Use With (Application) Water quality testing
Parameter Specifications Chlorine - free, high range
Quantity 50 Strips/Vial
Range 0-50-100-250-500-800 ppm
Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Type Single Factor Test Strip
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.