Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Insta-Test™ Water Test Strips
SDPSCI_ED

Catalog No. S66229 Shop All LaMotte Company Products
Change view
Click to view available options
Analytes:
Copper, Iron
Hardness
Nitrate, Nitrite
Total Chlorine
Total Chlorine, pH
pH
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analytes
S66229 Total Chlorine
S66231 pH
S66230 Hardness
S66234 Total Chlorine, pH
S66235 Nitrate, Nitrite
S66232 Copper, Iron
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S66229 Supplier LaMotte™ Supplier No. 2979
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform safe, quick and easy water quality checks with Insta-Test™ Water Test Strips.

  • Monitor water without having to use reagents or field kits.

Order Info

Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.

Specifications

Analytes Total Chlorine
Format Strip
For Use With (Application) Drinking water, food/beverage testing
Quantity 50 Pk.
Range 0 to 5 ppm (0, 0.5, 1, 3, 5 ppm)
Type Water Test Strip
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.