Learn More
Apexbio Technology LLC InstaBlue Protein Stain Solution, 1L. Cas: MFCD: MFCD00078482. For ultrafast protein gel staining
SDP
Catalog No. 501905499
Encompass_Preferred
InstaBlue Protein Stain Solution is a ready to use Coomassie protein stain for polyacrylamide gels. Its unique patented mechanism of action stains proteins in 5 minutes while leaving a clear background without the need to fix, wash or destain. InstaBlue Protein Stain Solution is formulated for safe use and easy disposal. InstaBlue Protein Stain Solution is very convenient for use and needs only simple one step protocol to finish the rapid coomassie blue staining of protein gels. Some premade and traditional home-made Coomassie R-250 protein stains can take 3 hours or more to fully stain gels. Because of InstaBlues protein specific staining which gives a clear background, improving the signal/noise ratio of the stain, InstaBlue Protein Stain Solution has high sensitivity and can detect protein levels as low as 5 ng per band when staining overnight. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5499 Supplier Apexbio Technology LLC Supplier No. B82261000
InstaBlue Protein Stain Solution is a ready to use Coomassie protein stain for polyacrylamide gels. Its unique patented mechanism of action stains proteins in 5 minutes while leaving a clear background without the need to fix, wash or destain. InstaBlue Protein Stain Solution is formulated for safe use and easy disposal. InstaBlue Protein Stain Solution is very convenient for use and needs only simple one step protocol to finish the rapid coomassie blue staining of protein gels. Some premade and traditional home-made Coomassie R-250 protein stains can take 3 hours or more to fully stain gels. Because of InstaBlues protein specific staining which gives a clear background, improving the signal/noise ratio of the stain, InstaBlue Protein Stain Solution has high sensitivity and can detect protein levels as low as 5 ng per band when staining overnight. Other sizes are available. Please inqury us for quote.