Learn More
Hach Company Installation Kit for 5500sc Silica/Phosphate Analyzers (for 5500sc analyzers)
Catalog No. NC3361128 Shop All Hach Company Products
Encompass
The installation kit for 5500sc Silica/Phosphate analyzers provides the components and basic tools required to install and commission a 5500sc analyzer. It supplies sample conditioning, mounting hardware, grounding, and hand tools to complete initial setup and protect instrument components during installation and first use.
  • Includes pressure regulator set to 4 psi.
  • Includes Y-strainer for sample protection.
  • Includes Torx drivers (T20, T25) for assembly.
  • Includes stir bar and tubing for sample lines.
  • Includes screws, connectors, and a sample valve for plumbing.
  • Includes grounding kit and cables for safe installation.
  • Includes bags, labels, and a plastic funnel for organization.
Catalog No. NC3361128 Supplier Hach Company Supplier No. 6783500
The installation kit for 5500sc Silica/Phosphate analyzers provides the components and basic tools required to install and commission a 5500sc analyzer. It supplies sample conditioning, mounting hardware, grounding, and hand tools to complete initial setup and protect instrument components during installation and first use.
  • Includes pressure regulator set to 4 psi.
  • Includes Y-strainer for sample protection.
  • Includes Torx drivers (T20, T25) for assembly.
  • Includes stir bar and tubing for sample lines.
  • Includes screws, connectors, and a sample valve for plumbing.
  • Includes grounding kit and cables for safe installation.
  • Includes bags, labels, and a plastic funnel for organization.