Learn More
MilliporeSigma Application-Pak Installation Kit
Catalog No. NC1688748
Encompass
EDS-Pak Cartridge installation and Conditioning Kit including the following reusable parts: Polyethylene ¼ Gaz F-Hose barb connection with O-ring, Polyethylene ¼ G F – ¼ Gaz F connectors for connection to Millipak
Catalog No. NC1688748 Supplier MilliporeSigma Supplier No. EDSKIT001
EDS-Pak Cartridge installation and Conditioning Kit including the following reusable parts: Polyethylene ¼ Gaz F-Hose barb connection with O-ring, Polyethylene ¼ G F – ¼ Gaz F connectors for connection to Millipak