Learn More
Julabo USA Inc Each, cooling coil for SW22/SW23 shaking water baths.
SDP
Catalog No. 507534182
Encompass_Preferred
Bath Accessories, Julabo, Cooling coil for retrofitting shaking water baths SW22/SW23. The cooling coil is screwed into the water bath through the drain tap. Once installed, the bath may be operated near or slightly below ambient temperature.
Catalog No. 50-753-4182 Supplier Julabo USA Inc Supplier No. 8970416
Bath Accessories, Julabo, Cooling coil for retrofitting shaking water baths SW22/SW23. The cooling coil is screwed into the water bath through the drain tap. Once installed, the bath may be operated near or slightly below ambient temperature.