Learn More
DWK LIFE SCIENCES™ WHEATON™ Installation Kit for WHEATON™ Bottom-Drive R2P 2.0 Roller Apparatus
Shop All DWK Life Sciences Products
Description
- Field installation options package for the Wheaton R2P 2.0 Bottom-Drive Roller Apparatus
- Includes battery backup to protect cells during transport, processing, and power outages
- Dual temperature sensors enable real-time temperature monitoring and deviation alarms
- Temperature displayed on touch screen with screen saver for quick confirmation of optimal operating conditions
- Rotation (belt break) alarm is already included with the R2P 2.0 unit
- Belt break alarm employs two magnetic sensors to ensure all main pulleys are rotating
Specifications
Specifications
| Product Type | Installation Kit |
| For Use With (Equipment) | Wheaton Bottom-drive R2P 2 Roller Apparatus |
| Includes | Automatic battery backup and dual temperature sensors/alarm |